ContactUsLinkComponent
One-Piece Rulers, Lufkin®, ORS Nasco
  300062-778
undefined
One-Piece Rulers, Lufkin®, ORS Nasco
  300062-778
 :  182-626FT
  • Description:
    Rule
  • Length:
    6 ft.
  • Material:
    Steel
  • Cat. No.:
    300062-778
  • Width:
    1¼"
  • Supplier no.:
    182-626FT
Bold markings, deeply etched and permanent are black filled.
  • Heavier than on machine-divided rules
  • Large graduations, marked both sides, both edges
  • Upper edge graduated in ¹/₈ths, lower edge in ¹/₁₆ths
  • Opposite sides measure from opposite ends