Steel Protractors, L.S. Starrett
Catalog # 300046-746
Supplier: Safety & Industrial Supplies
Specifications
- Cat. No.:300046-746
- Supplier no.:681-50127
Specifications
About this item
Steel protector 6"
- Double graduations from 0-180° in opposite directions permitting the direct reading of angles and supplementary angles
- The back of the tool is flat for ease of use
- The blade can be locked firmly at any angle by the lock nut
- Satin chrome finish for ease of reading and resistance to rust
- Rectangle head
For setting bevels, transferring angles, small squaring tasks, checking cutter clearances within certain limits, and other applications.