Digital/Fraction Electronic Calipers, General Tools
Supplier: Safety & Industrial Supplies
Digital/Fraction Electronic Calipers, General Tools
Supplier: Safety & Industrial Supplies
Certificates
About this item
- Large, easy to read LCD numbers
- On/Off and zero button
- Pocket clip
2 Options Available Below
Product Details & Documents
- Large, easy to read LCD numbers
- On/Off and zero button
- Pocket clip
Specifications for Product Family
Specifications
- Catalog No:
- 300052-450
- 300078-866
- Description:
- 300052-450
- 300078-866
- Body Material:
- Hardened stainless steel
- Hardened stainless steel
- Display Type:
- Digital LCD
- Digital LCD
- Includes:
- Carrying Case
- Carrying Case
- Lower Jaw Type:
- Plain Jaw
- Plain Jaw
- Measurement Type:
- Depth, Inside, Outside, Step
- Depth, Inside, Outside, Step
- Resolution (Length):
- 0.000500 in, 0.010000 mm
- 0.000500 in, 0.010000 mm
- Type:
- Plain Caliper
- Upper Jaw Type:
- Plain Jaw
- Plain Jaw
Specifications
Product Family Options
Product Information
- ResolutionMeasurement rangeAvailabilityPrice
- 1/8,1/4, 1/2, 1/16, 1/32 & 1/64"0" [Min], 0 mm [Min], 6" [Max], 150 mm [Max]Log in for availabilityEach$115.21Supplier Number: 318-147Specifications:
More
Specifications
Cat. No.300052-450Description300052-450Body MaterialHardened stainless steelDisplay TypeDigital LCDIncludesCarrying CaseLower Jaw TypePlain JawMeasurement TypeDepth, Inside, Outside, StepResolution (Length)0.000500 in, 0.010000 mmTypePlain CaliperUpper Jaw TypePlain Jaw - 1/8,1/4, 1/2, 1/16, 1/32 & 1/64"1" [Min], 0 mm [Min], 3" [Max], 150 mm [Max]Log in for availabilityEach$37.01Supplier Number: 318-1433Specifications:
More
Specifications
Cat. No.300078-866Description300078-866Body MaterialHardened stainless steelDisplay TypeDigital LCDIncludesCarrying CaseLower Jaw TypePlain JawMeasurement TypeDepth, Inside, Outside, StepResolution (Length)0.000500 in, 0.010000 mmTypeUpper Jaw TypePlain Jaw