Professional Lysol® Brand III Disinfectant Spray, Reckitt Benckiser
Supplier: Avantor
Certificates
About this item
- Highly effective against TB, MRSA, HIV-1 (AIDS virus), poliovirus Type 1, hepatitis A, and norovirus
- Tuberculocidal, virucidal, fungicidal, bactericidal
- Eliminates odors and prevents the growth of mold and mildew
- Contains no CFCs
- Aerosol can
Frequently Bought Together