Hollow Punch Tool Kits, 27 Piece, Mayhew™ Tools, ORS Nasco
300005-562
- Material:Alloy steel
- Type:Punch Set
- Cat. No.:300005-562
- Weight:6.06 lbs.
- Includes:Mandrin, handle, case
- Finish:Black oxide
- Supplier no.:479-66002
- Tip type:Round
- Cutting diameter:⅛, ³⁄₁₆, ¼, ⁵⁄₁₆, ⅜, ⁷⁄₁₆, ½, ⁹⁄₁₆, ⅝, ¾, ⅞, 1, 1¹⁄₁₆, 1⅛, 1³⁄₁₆, 1¼, 1⁵⁄₁₆, 1⅜, 1⁷⁄₁₆, 1½, 1⅝, 1¹¹⁄₁₆, 1¾, 1⅞, 2, ⅛" (min.), 2 (max.)"