Torqueplus™ Protoblack™ Metric 12-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
- TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
- Reduces risk of fastener rounding
- Applies more torque to fastener
- Unique arc design increases surface contact
- Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
- Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Specifications
- Catalog No:
- 300023-662
- 300023-590
- 300023-582
- 300023-698
- 300023-560
- 300023-736
- 300023-570
- 300023-652
- 300023-630
- 300023-686
- 300023-620
- 300023-742
- 300023-708
- 300023-674
- 300023-718
- 300023-640
- 300023-726
- 300023-604
- Type:
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Head Angle:
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- Offset Angle:
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- Opening Type:
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box