Torqueplus™ Protoblack™ 12-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
- TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
- Reduces risk of fastener rounding
- Applies more torque to fastener
- Unique arc design increases surface contact
- Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
- Improves grip on worn, rounded fasteners
- Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
- For access to areas where sockets and attachments will not fit or where greater leverage is required
Specifications
- Catalog No:
- 300030-798
- 300030-794
- 300023-614
- 300030-776
- 300030-780
- 300030-788
- 300023-802
- 300030-778
- 300030-782
- 300030-786
- 300030-774
- 300030-790
- 300030-784
- 300023-798
- 300030-800
- 300030-802
- 300030-796
- 300023-810
- 300030-804
- 300023-812
- 300023-806
- 300023-788
- 300023-794
- Type:
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Head Angle:
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- Offset Angle:
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- Opening Type:
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box