Torqueplus™ Flare Nut Crowfoot Wrench Drive Bits, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
- Side lock hole
- Reduces risk of fastener rounding
- Applies more torque to fastener
- High strength, thin wall design
Specifications
- Catalog No:
- 300025-688
- 300025-686
- 300025-692
- Type:
- Crowfoot
- Crowfoot
- Crowfoot
- Drive Size:
- 3/8 in
- 3/8 in
- 3/8 in
- Drive Type:
- Square
- Square
- Square
- Opening Type:
- Flare
- Flare
- Flare