ContactUsLinkComponent
Torqueplus™ Combination Flare Nut Wrenches, Proto®, ORS Nasco
Torqueplus™ Combination Flare Nut Wrenches, Proto®, ORS Nasco
Certificates
undefined
Torqueplus™ Combination Flare Nut Wrenches, Proto®, ORS Nasco
Specifications
  • Catalog No:
  • 300024-802
  • 300024-806
  • Type:
  • Flare Nut
  • Flare Nut
  • Head Angle:
  • 15° Open Side, 30° Flare Side
  • 15° Open Side, 30° Flare Side
  • Offset Angle:
  • 15° Flare Side
  • 15° Flare Side
  • Opening Type:
  • Open/Flare
  • Open/Flare