Torqueplus™ 6-Point Short Combination Wrenches, Proto®, ORS Nasco
- Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
- Applies more torque to fastener
- Reduces risk of fastener rounding
- Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
- TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
- Unique arc design increases surface contact
- Catalog No:
- 300023-588
- 300023-600
- Type:
- Combination
- Combination
- Head Angle:
- 15° Open Side
- 15° Open Side
- Offset Angle:
- 15° Box Side
- 15° Box Side
- Opening Type:
- Open/Box
- Open/Box
Frequently Bought Together