Torqueplus™ 12-Point Metric Combination Wrenches - Satin Finish, Proto®, ORS Nasco
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
- Reduces risk of fastener rounding
- Applies more torque to fastener
- Unique arc design increases surface contact
- Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
- Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
- Catalog No:
- 300002-730
- 300023-696
- 300002-734
- 300023-672
- 300023-558
- 300002-712
- 300023-568
- 300023-768
- 300023-580
- 300023-618
- 300023-706
- 300023-780
- 300023-716
- 300023-792
- 300023-724
- 300023-650
- 300023-756
- 300023-602
- 300023-638
- 300023-786
- 300023-764
- 300002-708
- 300023-660
- 300023-734
- 300023-740
- 300023-790
- 300023-750
- 300023-684
- 300023-760
- Type:
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Combination
- Head Angle:
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- 15° Open Side
- Offset Angle:
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- 15° Box Side
- Opening Type:
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
- Open/Box
Frequently Bought Together