Torqueplus™ 12-Point Combination Flare Nut Wrenches, Proto®, ORS Nasco
Applies more torque to fastener.
- Modified square open end provides better fastener grip
- Unique arc design increases surface contact
- Increased torque applied to fastener with fewer rounded edges
- Improves grip on worn, rounded fasteners
- Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
- Catalog No:
- 300024-826
- 300024-814
- 300024-818
- Type:
- Flare Nut
- Flare Nut
- Flare Nut
- Head Angle:
- 15° Flare Side, 15° Open Side
- 15° Flare Side, 15° Open Side
- 15° Flare Side, 15° Open Side
- Opening Type:
- Open/Flare
- Open/Flare
- Open/Flare
Frequently Bought Together