Super Heavy-Duty Offset Slugging Wrenches, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
- Heavy box walls provide greater torque transmission
- Box opening remains on fastener when wrench is struck
- High strength rectangular design withstands heavy sledging
- Straight handle resists springing for easier nut turning
- For close quarter work where hammer or sledge is required to loosen frozen nuts or set large fasteners
Specifications
- Catalog No:
- 300028-718
- 300028-750
- 300028-742
- 300028-758
- 300028-754
- 300028-762
- 300028-764
- Type:
- Striking
- Striking
- Striking
- Striking
- Striking
- Striking
- Striking
- Offset Angle:
- 45° Box Side
- 45° Box Side
- 45° Box Side
- 45° Box Side
- 45° Box Side
- 45° Box Side
- 45° Box Side
- Opening Type:
- Box
- Box
- Box
- Box
- Box
- Box
- Box