Proto® Open End Wrench Set, 10 Piece, Stanley
Catalog # 300024-431
Supplier: Safety & Industrial Supplies
Specifications
- Description:10 Piece Open End Wrench Set
- Cat. No.:300024-431
Specifications
About this item
Increased torque applied to fastener with fewer rounded edges
- Unique arc design increases surface contact
- Forged alloy steel construction
- Flat handle
- 15° head angle
- Measurements provided in inches
Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury.
Delivery: Set includes (1) 1/4 x 5/16", (1) 5/16 x 3/8", (1) 7/16 x 1/2", (1) 1/2 x 9/16", (1) 9/16 x 5/8", (1) 5/8 x 3/4", (1) 11/16 x 3/4", (1) 13/16 x 7/8", (1) 15/16 x 1", (1) 11/16 x 11/8", and tool roll.