Telescoping MegaMag® Magnetic Pick-Up Tools, Ullman, ORS Nasco
Catalog # 300005-130
Supplier: Safety & Industrial Supplies
Specifications
- Length:7⅛"
- Handle Material:Stainless Steel
- Cat. No.:300005-130
- Supplier no.:758-GM-2
- Handle type:Cushion Grip
Specifications
About this item
Tool capacity: 16 lbs. [Max]
- Comfortable textured cushion pistol grip will not slip in wet or oily hands
- It's sides will not attach to unwanted metal surfaces
- It can be extended upto in 30¼"
- Mirror with metal base mounts to MegaMag® in seconds to bring hidden areas into full view