ContactUsLinkComponent
Hook and Pick Sets, 6 Piece, Ullman, ORS Nasco
Hook and Pick Sets, 6 Piece, Ullman, ORS Nasco
Catalog # 300005-126
undefined
Hook and Pick Sets, 6 Piece, Ullman, ORS Nasco
Catalog # 300005-126
Supplier Number:  758-CHP6-L
Specifications
  • Material:
    High Carbon Steel
  • Cat. No.:
    300005-126
  • Includes:
    Combination Pick, Hook Pick, Straight Pick, 90° Pick, Complex Pick, Double Angle Pick
  • Supplier no.:
    758-CHP6-L
Specifications
About this item
Ideal for separating wires, removing small fuses, "O" rings, retrieving washers and loose parts too small to pick up with fingers
Each pick has a cushioned textured grip that will not slip in wet or oily hands.