Tube Cutter Wheels, Ridgid®, ORS Nasco
Catalog # 300034-176
Supplier: Safety & Industrial Supplies
Specifications
- Cat. No.:300034-176
- Weight:0.02 lbs.
- Supplier no.:632-74720
Specifications
About this item
Cutter wheels for PB, PE, PP, standard and heavy wall tubes.
- Blade Exp: 0.286
- Fits to 152-P, 153-P model cutters