ContactUsLinkComponent
Tube and Hose Shears, Imperial Stride Tool, ORS Nasco
  300004-960
undefined
Tube and Hose Shears, Imperial Stride Tool, ORS Nasco
  300004-960
 :  389-307-FP
  • Material:
    Steel
  • Cat. No.:
    300004-960
  • Weight:
    1 lb.
  • Supplier no.:
    389-307-FP
Strong shears are designed to cut through plastic tubing and non-metal reinforced hose.
  • Spring loaded for safety
  • Pop-Up counter display
  • Replaceable hardened steel blade
Shears close automatically, eliminating worry about the tool being left with blades exposed.