ContactUsLinkComponent
PVC Cutters, Greenlee®, ORS Nasco
PVC Cutters, Greenlee®, ORS Nasco
Catalog # 300019-830
undefined
PVC Cutters, Greenlee®, ORS Nasco
Catalog # 300019-830
Supplier Number:  332-864
Specifications
  • Cat. No.:
    300019-830
  • Cutting capacity:
    1¼"
  • Weight:
    0.80 lbs.
  • Supplier no.:
    332-864
Specifications
About this item
Cutters to cut plastic pipe, PVC and rubber hose.
  • Stainless Steel blade
  • Ratchet type