ContactUsLinkComponent
2000 High-Leverage Diagonal Cutter Plier, Klein Tools, ORS Nasco
2000 High-Leverage Diagonal Cutter Plier, Klein Tools, ORS Nasco
Catalog # 300005-554
undefined
2000 High-Leverage Diagonal Cutter Plier, Klein Tools, ORS Nasco
Catalog # 300005-554
Supplier Number:  409-D2000-48
Specifications
  • Color:
    Royal Blue
  • Length:
    20.3 cm (8")
  • Material:
    Alloy steel
  • Cat. No.:
    300005-554
  • Supplier no.:
    409-D2000-48
Specifications
About this item
Ultra-durable "2000 Series" cutting pliers for ACSR, bolts, nails, and most hardened wire.
  • Precision machined, mating surfaces and design dimensions to extremely close tolerances
  • Pliers halves are matched side to side, jaw to jaw, and hot riveted
  • Drop forged, hardened, and tempered construction
  • Angled head design for easy work in confined spaces