ContactUsLinkComponent
12 Pc. Swiss Pattern Needle File Sets, General Tools
12 Pc. Swiss Pattern Needle File Sets, General Tools
Catalog # 300004-260
undefined
12 Pc. Swiss Pattern Needle File Sets, General Tools
Catalog # 300004-260
Supplier Number:  318-707475
Specifications
  • Description:
    Set including (1) Warding (Flat) File, (1) Round File, (1) Three Square File, (1) Equaling (Mill) File, (1) Square File, (1) Half Round File, (1) Crossing File, (1) Oval File, (1) Knife File, (1) Barrette File, (1) Slitting File, (1) Joint Edge Round File, Handle, Vinyl Pouch
  • Length:
    5½"
  • Material:
    Alloy steel
  • Cat. No.:
    300004-260
  • Supplier no.:
    318-707475
Specifications
About this item
Needle file set incl. a vinyl pouch and 12 files.
  • Ergonomic handle, with contoured fingertip swivel, allows greater pressure to be applied to blade
  • Precision locking screw chuck holds files securely in place
Shank diameter: 1/8"
Files included:
(1) Warding (Flat) File
(1) Round File
(1) Three Square File
(1) Equaling (Mill) File
(1) Square File
(1) Half Round File
(1) Crossing File
(1) Oval File
(1) Knife File
(1) Barrette File
(1) Slitting File
(1) Joint Edge Round File