BD Falcon™ SWUBE™ Pinworm Paddle Collection Device, BD Diagnostics
Catalog # 90001-060
Supplier: Avantor
Specifications
- Description:Pinworm Paddle Collection Device, Sterile
- Cat. No.:90001-060
- Supplier no.:220120
Specifications
About this item
A clear plastic paddle housed inside a 17x100 mm plastic tube.
Specially designed for the collection and transport of suspected pinworm samples. It may also be used for spore and pollen collection. The paddle features a nontoxic adhesive coating that holds the specimen. The paddle is designed to fit directly onto a microscope stage for easy specimen examination.