Specifications
- Flow rate:15 drops/mL
- Length:267 cm (105")
- Cat. No.:95037-776
- Supplier no.:NF3482
Specifications
About this item
IV administration set includes a universal spike, roller clamp, slide clamp, SAFELINE® valve injection site (26") above distal end, and SPIN-LOCK® connector. Priming volume: 19 mL.