- Length:76.2 cm (30")
- Cat. No.:72220-000
- Supplier no.:513501
Transfer set with weighted sinker and tubing clip for use with MULTI-AD® pump or for manual activation use when dispensing fluids from an open container
The set is DEHP-free and latex-free. Includes a 10cc luer lock syringe with adjustable automatic spring return and dual check valve.