ContactUsLinkComponent
Total Protein Extraction Kit, TPE™, G-Biosciences
Total Protein Extraction Kit, TPE™, G-Biosciences
Catalog # 82021-366
Supplier:  G-Biosciences
CAS Number:  
Total Protein Extraction Kit, TPE™, G-Biosciences
Catalog # 82021-366
Supplier:  G-Biosciences
Supplier Number:  786-225
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Total Protein Extraction (TPE™) Kit
  • Size:
    50 preps
  • Cat. No.:
    82021-366
  • Supplier no.:
    786-225
Specifications
About this item
G-Biosciences' Total Protein Extraction (TPE™) Kit is a universal lysis system for the solubilization of total proteins from animal, plant, yeast, bacteria, and other biological samples
The TPE™ kit provides a two component protocol that eliminates clump formation, protein loss, and other problems associated with total protein extraction procedures. Samples are ground in the buffer provided and then heated to solubilize the total protein.
The TPE™ kit is based on optimized concentration of Tris and SDS and is suitable for running denaturing electrophoresis and other downstream applications. The kit is supplied with sufficient reagents for 50 x 250mg preparations.
The kit contains ready-to-use buffers for extracting total protein from cells and tissues for SDS-PAGE analysis.
Ordering information: Accessories are available.