Order Entry
United States
Orders LinkContactUsLinkComponent
Waypoint® Flashlights, Streamlight®

Waypoint® Flashlights, Streamlight®

Yellow, 6.75"L x 7.1"H, C4 LED
Catalog # 300053-210
Supplier #:  683-44900
Waypoint® Flashlights, Streamlight®
Catalog # 300053-210
Waypoint® Flashlights, Streamlight®
Waypoint® Flashlights, Streamlight®

Yellow, 6.75"L x 7.1"H, C4 LED

Price
$152.46
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Flashlight with high-impact polycarbonate body.

  • Hand-held, pistol grip spotlight features a super bright C4 LED
  • High, low, and emergency signal modes
  • Beam concentration: 115000 cp, 10000 cp
  • 50000 hour lamp life
  • For use with batteries or with power cord.

4 Size C Alkaline required. 12V DC power cord and high strength lanyard included.

Specifications
  • Color:
    Yellow
  • Dimensions:
    6.75"L x 7.1"H
  • Bulb type:
    C4 LED
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.