ContactUsLinkComponent
Spotcheck® Kit, Magnaflux
Spotcheck® Kit, Magnaflux
  300008-507
undefined
Spotcheck® Kit, Magnaflux
  300008-507
 :  387-01-5920-48
  • Description:
    Spotcheck® kit. Includes: (2) cans of SKD-S2 developer, (2) cans of SKL-SP1 penetrant, (4) cans of SKC-S cleaner/remover, instructions, paint marker, portable plastic case, scrub hand towels, wiping cloth
  • Cat. No.:
    300008-507
  • Supplier no.:
    387-01-5920-48
Spotcheck® visible penetrant kits quickly locate surface cracks.
  • Easy to transport for field inspections
  • No UV light required because of vivid red indications