Precision Clean Multi-Purpose Cleaner/Degreaser, LPS®
Catalog # 300050-626
Supplier: Safety & Industrial Supplies
Specifications
- Size:28 oz.
- Cat. No.:300050-626
- Packaging:Trigger spray bottle
- Supplier no.:428-02728
Specifications
About this item
Water-based biodegradable formula removes tough, stubborn stains and soils such as grease, oil, sludge, soot, ink, coffee, and more.
- pH: 12.5–13.0
- Does not contain chlorinated or petroleum solvents, bleach, ammonia, abrasives or butyl
- Contains rust and corrosive inhibitor to prevent flash rusting of metal surfaces
- Safe on all surfaces except magnesium
- Nonflammable