Order Entry
United States
ContactUsLinkComponent
Fluoresbrite® YG Microspheres, Calibration Grade 6 µm, Polysciences
Fluoresbrite® YG Microspheres, Calibration Grade 6 µm, Polysciences
Catalog # 87001-906
Supplier:  Polysciences Inc
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
undefined
Fluoresbrite® YG Microspheres, Calibration Grade 6 µm, Polysciences
Catalog # 87001-906
Supplier:  Polysciences Inc
Supplier Number:  18862-1
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Microspheres Fluoresbrite® YG, Calibration Grade 6µm
  • Size:
    1 mL
  • Environmentally Preferable:
  • Cat. No.:
    87001-906
  • Supplier no.:
    18862-1

Specifications

About this item

For flow cytometry standards

A special grade of Yellow Green Fluoresbrite® beads has been evaluated for both particle diameter and uniformity of dye distribution. Packaged in an economical suspension of 2.5% solids in water