ContactUsLinkComponent
PhosphoQuant™, G-Biosciences
PhosphoQuant™, G-Biosciences
Catalog # 82022-698
Supplier:  G-Biosciences
CAS Number:  
undefined
PhosphoQuant™, G-Biosciences
Catalog # 82022-698
Supplier:  G-Biosciences
Supplier Number:  786-256
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PhosphoQuant™
  • Cat. No.:
    82022-698
  • Supplier no.:
    786-256
Specifications
About this item
PhosphoQuant™ is specifically designed for quick and reliable determination of whether a purified protein is phosphorylated and the extent of phosphorylation
The assay is based on the alkaline hydrolysis of phosphates from seryl and threonyl residues in phosphoproteins and the subsequent quantification of the released phosphate with a Molybdate dye.
Ordering information: Accessories are available.