ContactUsLinkComponent
Blood Alcohol Collection and Transport Kit, Therapak®
Blood Alcohol Collection and Transport Kit, Therapak®
Catalog # 89171-034
Supplier:  Therapak, LLC
undefined
Blood Alcohol Collection and Transport Kit, Therapak®
Catalog # 89171-034
Supplier:  Therapak, LLC
Supplier Number:  38107
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Blood Alcohol Collection and Transport Kit
  • Environmentally Preferable:
  • Cat. No.:
    89171-034
  • Supplier no.:
    38107
Specifications
About this item
Kit contains all the necessary components for the collection and transport of blood samples for alcohol testing
Included are two 16x100mm evacuated blood tubes with sodium fluoride – potassium oxalate preservative, a blood draw needle, needle holder, iodine prep pad, plastic tube holder, two evidence seals, box seal, transport bag with absorbent material, marking labels, and transport box
Certifications: Compliant for shipping by ground or air in accordance with Exempt Human Specimen shipping regulations.