Clear RTV Silicone Adhesive Sealants, Permatex®
Supplier: Safety & Industrial Supplies
Certificates
About this item
Waterproof and flexible sealant may be applied to horizontal or vertical surfaces.
- Seals, bonds, repairs, mends, and secures
- Applicable materials include glass, metal, plastics, fabric, vinyl, weather stripping, and vinyl tops
- –75 to 400°F temperature range
- Offers resistance to oil, water, weather, vibration, and grease
Frequently Bought Together