5 Minute® Epoxy Gel, Devcon
Supplier: Safety & Industrial Supplies
Jump to:
About this item
Thixotropic, nonmigrating gel adhesive with excellent gap filling properties.
- High tensile strength
- Good solvent resistance
- Fills gaps to 0.25"
Non-sagging adhesive is ideal for use on vertical surfaces.
2 Options Available Below
Product Details & Documents
Thixotropic, nonmigrating gel adhesive with excellent gap filling properties.
- High tensile strength
- Good solvent resistance
- Fills gaps to 0.25"
Non-sagging adhesive is ideal for use on vertical surfaces.
Documents
Product Family Options
Product Information
- ColorVolumeAvailabilityPrice
- 300050-520Epoxy gel in Dev-Pak™Supplier Number: 230-14265
- 300050-778Epoxy gel in dual syringe tubeSupplier Number: 230-14240