About this item
Monitor the performance of various cancer antigen tests on automated instruments with liquid assayed, multi-analyte MAS™ T-Marker Controls
Liquid, ready to use. No additional preparation required. Human serum matrix. Includes CA 15-3, CA 19-9, CA 72-4, CA-125, CA 27-29. Assayed values available for most major analyzers.
Analytes: ACTH, AFP, Aldosterone, CA 15-31, CA 19-91, CA 72-41, CA-125, CA-29, Calcitonin, CEA, Cortisol, Estradiol, Ferritin, Gastrin, hCG, ?-hCG, Insulin, ?-2-microglobulin, PAP, Prolactin, PSA, PSA (free), Thryoglobulin.
These controls contain more 24 analytes, including a complete offering of cancer-associated antigens.