Streck Cell Preservative™ Cell Stabilization Reagents, Streck
Supplier: Streck Laboratories
Certificates
About this item
Easy-to-use liquid preservatives stabilize cell morphology and preserve cellular antigen expression
Can be used with bone marrow aspirates, surgical tissue samples, fine needle aspirates, or peripheral blood. Samples collected in this preservative remain stable for up to seven days.
Frequently Bought Together