ICON® 20 hCG Pregnancy Test, Beckman Coulter®
89038-224
: Hemocue
:
- Description:ICON® 20 hCG Pregnancy Test
- Cat. No.:89038-224
- Supplier no.:395097A
Rapid test for detecting hCG qualitatively.
- Qualitative
- Sensitive (Serum 10 mIU/mL, Urine 20 mIU/mL )
- Solid-phase chromatography
- Detect hCG
Uses a solid-phase chromatographic immunoassay technology to selectively detect elevated levels of hCG in serum or urine. Simple, one-step process for greater reliability and higher sensitivity. Built-in positive and negative controls.
: Urine is CLIA waived; serum is moderately complex.
: Kit contents include 25 test devices and 25 disposable droppers.