Sickle-Chex Controls, Streck
Supplier: Streck Laboratories
Certificates
About this item
Positive and negative whole blood controls are for use with sickle cell screening tests and hemoglobim electrophoresis
Do not need to be reconstituted prior to use, which allows the controls to be utilized like a patient sample. Packaged in 2.5mL squeeze dropper vials for easy dispensing. Controls have a 100-day open-vial stability and 6-month closed-vial stability
Frequently Bought Together