A1c-Cellular® Whole Blood Cellular Controls, Streck
Supplier: Streck Laboratories
Certificates
About this item
Controls test the entire HbA1c procedure, incluing the lysing of the red blood cell, which ensures the entire system, instrument and reagents are working properly
The controls have an open-vial stability of 30 days, and a closed-vial stability of 6 months when stored at 2 to 10°C. Available in plastic vials with pierceable caps.
Frequently Bought Together