ContactUsLinkComponent
Hypodermic Syringes with Needles, Terumo®
Hypodermic Syringes with Needles, Terumo®
Catalog # 15003-046
Supplier:  Terumo
Hypodermic Syringes with Needles, Terumo®
Catalog # 15003-046
Supplier:  Terumo
Supplier Number:  SS-03L2138
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    3 cc
  • Environmentally Preferable:
  • Cat. No.:
    15003-046
  • Needle length:
    3.81 cm (1½”)
  • Gauge:
    21
  • Graduations:
    0.1 cc
  • Supplier no.:
    SS-03L2138
Specifications
About this item
Syringes come with ultra-thin wall, ultra-sharp needles attached
The needles are precisely honed and beveled for ease of penetration. The latex-free clear syringes have bold scale markings allowing quick visual inspection for precise dosage. An integrated plunger stop helps prevent the plunger from being accidentally pulled out