MULTI-CORE™ Buffer Pack, Promega
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Universal restriction enzyme buffer. Supports double-digests with many restriction enzymes.
- Convenient Aliquots of the Promega Universal Restriction Enzyme 10X Buffer
- Enables digestion of DNA with more than one enzyme in a single reaction
- Only modest adjustments in the amount of enzyme used will ensure complete multiple digestions
The MULTI-CORE™ Buffer is formulated to provide simple buffering conditions for performing multiple digestions.
MULTI-CORE™ Buffer (1X) = 25mM Tris-acetate (pH 7.5 at 37°C), 100mM potassium acetate, 10mM magnesium acetate, 1mM DTT.
Many Promega restriction enzymes have between 50% and 100% activity in reactions using this buffer.
Caution: For laboratory use. Outside of the United States, this product is intended for research use only unless otherwise stated.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:MULTI-CORE™ Buffer Pack
- Size:3 × 1 ml
- Cat. No.:PAR9991
- Storage Temperature:–30...–10 °C
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



