Order Entry
United States
ContactUsLinkComponent
Foam Biopsy Pads, BIO PLAS
Foam Biopsy Pads, BIO PLAS
Catalog # 21150-088
Supplier:  Bio Plas
undefined
Foam Biopsy Pads, BIO PLAS
Catalog # 21150-088
Supplier:  Bio Plas
Supplier Number:  6120

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Color:
    Blue
  • Cat. No.:
    21150-088
  • For use with:
    1 x 1 1/4" Cassettes
  • Supplier no.:
    6120

Specifications

About this item

Pads are fabricated from cellular polyester plastic foam with air pockets throughout to allow for adequate fluid movement around the tissue