Order Entry
United States
Orders LinkContactUsLinkComponent
Thallium(I) acetate 99%

Thallium(I) acetate 99%

Toxic
Health
Environment
Marketsource
Thallium(I) acetate 99%
Supplier:  
Marketsource
Certificates
Thallium(I) acetate 99%

0 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid thallium (I) salt, Thallous acetate
  • CAS Number:563-68-8
  • Formula:C₂H₃O₂Tl
  • MDL Number:MFCD00013045
  • Molecular Weight:263.43 g/mol
  • Merck Index:12,09392
  • Boiling Pt:1457 °C (1013 hPa)
  • Melting Pt:128…130 °C
  • Density:3.68 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-257-5
  • UN:1707
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥98.5 %
  • Titration with NaOH:
    ≥96.0 %
Product Family Options