Order Entry
United States
Orders LinkContactUsLinkComponent
Agar, powder, pure

Agar, powder, pure

Marketsource
Agar, powder, pure
Supplier:  
Marketsource
Certificates
Agar, powder, pure

0 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to cream to light yellow to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤20.0 % (1 g, 105 °C, 5 h)
  • Heavy metals:
    ≤20 ppm
  • Gel strength:
    500 to 1000 g/cm² (1.5 % at 20 °C)
  • Ash:
    ≤6 %
  • Microbial contamination:
    ≤1000 /g (Total plate count)
  • pH:
    5.7 to 7.5 (1.5 % in water)
Product Family Options
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.