Order Entry
United States
Orders LinkContactUsLinkComponent
 

Methyl laurate 98+%

Environment
Marketsource
Methyl laurate 98+%
Supplier:  
Marketsource
Certificates
Methyl laurate 98+%

0 Options Available Below

Product Details & Documents
Synonyms:  Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
  • CAS Number:111-82-0
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00008966
  • Molecular Weight:214.35 g/mol
  • Boiling Pt:262 °C (766 mmHg)
  • Melting Pt:4…5 °C
  • Density:0.87 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:203-911-3
  • UN:3082
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4290 to 1.4340 (20 °C, 589 nm)
  • GC:
    ≥98.0 %
Product Family Options
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.