(S)-(+)-Mandelic acid 99+%
Marketsource
About this item
Synonyms: L(+)-Mandelic acid, (S)-(+)-2-Hydroxy-2-phenylacetic acid, L(+)-2-Hydroxy-2-phenylacetic acid, (S)-2-Hydroxy-2-phenylacetic acid
- CAS Number:17199-29-0
- Formula:C₈H₈O₃
- MDL Number:MFCD00004495
- Molecular Weight:152.15 g/mol
- Boiling Pt:322 °C (1013 hPa)
- Melting Pt:130…135 °C
- Density:1.349 g/cm³ (20 °C)
- Flash Pt:>190 °C
- EINECS:241-240-8
0 Options Available Below
Product Details & Documents
Synonyms: L(+)-Mandelic acid, (S)-(+)-2-Hydroxy-2-phenylacetic acid, L(+)-2-Hydroxy-2-phenylacetic acid, (S)-2-Hydroxy-2-phenylacetic acid
- CAS Number:17199-29-0
- Formula:C₈H₈O₃
- MDL Number:MFCD00004495
- Molecular Weight:152.15 g/mol
- Boiling Pt:322 °C (1013 hPa)
- Melting Pt:130…135 °C
- Density:1.349 g/cm³ (20 °C)
- Flash Pt:>190 °C
- EINECS:241-240-8
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:130 to 135 °C
- Specific optical rotation:+152° to +158° (20 °C, 589 nm) (c=2, H2O)
- Titration with NaOH:≥99.0 %
- Chloride (Cl):≤0.1 ppm
- Nitrate (NO3):≤0.2 ppm
- Phosphate (PO4):≤0.05 ppm
- Sulfate (SO4):≤1 ppm
- Silicate (as SiO2):≤0.01 ppm
- Aluminium (Al):≤0.02 ppm
- Ammonium (NH4):≤0.01 ppm
- Antimony (Sb):≤0.01 ppm
- Barium (Ba):≤0.01 ppm
- Bismuth (Bi):≤0.01 ppm
- Boron (B):≤0.01 ppm
- Cadmium (Cd):≤0.01 ppm
- Calcium (Ca):≤0.1 ppm
- Chromium (Cr):≤0.01 ppm
- Cobalt (Co):≤0.01 ppm
- Copper (Cu):≤0.01 ppm
- Gold (Au):≤0.01 ppm
- Iron (Fe):≤0.01 ppm
- Lithium (Li):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Manganese (Mn):≤0.01 ppm
- Mercury (Hg):≤0.05 ppm
- Molybdenum (Mo):≤0.01 ppm
- Nickel (Ni):≤0.01 ppm
- Potassium (K):≤0.05 ppm
- Selenium (Se):≤0.01 ppm
- Silver (Ag):≤0.01 ppm
- Sodium (Na):≤0.1 ppm
- Strontium (Sr):≤0.05 ppm
- Tin (Sn):≤0.01 ppm
- Titanium (Ti):≤0.01 ppm
- Vanadium (V):≤0.01 ppm
- Zinc (Zn):≤0.05 ppm
- Zirconium (Zr):≤0.01 ppm
- Heavy metals (as Pb):≤0.01 ppm
- KMnO4-reducing substance:Passes test
- Conductivity:≤6 µS/cm



