Order Entry
United States
Orders LinkContactUsLinkComponent
 

Bis(2-ethoxyethyl)ether 99%

Bis(2-ethoxyethyl)ether 99%
Certificates
Bis(2-ethoxyethyl)ether 99%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 112-36-7 Molecular Formula: C8H18O3 Formula Weight: 162.23 Flash Point: 71°(160°F) Refractive Index: 1.4120 Odor: Weak Melting Point: -44° Boiling Point: 188-190° Beilstein Reference: 1699259 Merck Reference: 14,3118 MDL No.: MFCD00009254

Synonyms:  2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether
  • CAS Number:112-36-7
  • Formula:C₈H₁₈O₃
  • MDL Number:MFCD00009254
  • Molecular Weight:162.23 g/mol
  • Merck Index:14,3118
  • Boiling Pt:188…190 °C
  • Melting Pt:–44 °C
  • Density:0.909
  • Flash Pt:71 °C(160 °F)
  • Storage Temperature:Ambient
  • EINECS:203-963-7
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.