Order Entry
United States
ContactUsLinkComponent
 
Methyl orange 0.1% (w/v) in aqueous solution
Catalog # AA38695-AP
CAS Number:  
Methyl orange 0.1% (w/v) in aqueous solution
Catalog # AA38695-AP
Supplier Number:  038695-AP
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Size:
    500 mL
  • Cat. No.:
    AA38695-AP
  • Supplier no.:
    38695-AP

Specifications

About this item

CAS Number:547-58-0
 :
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502
 :327.34 g/mol
Density:0.987
Storage Temperature:Ambient
EINECS:208-925-3