
Potassium hydroxide 10% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Density:1.087
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together