Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3% ACS
:109-86-4
:
:C₃H₈O₂
:MFCD00002867
:76.1 g/mol
:14,6038
:124…125 °C
:–85 °C
:0.965
:46 °C (115 °F)
:Ambient
:203-713-7
:1188
:3,III
:GHS08
:GHS02
:GHS07
- Color (APHA):10
- Titratable acid:0.002meq/gmeq/g
- H2O:0.001%
Frequently Bought Together