Order Entry
United States
ContactUsLinkComponent
Hafnium ≥99.6% (trace metals basis excluding Zr), powder -60+325 mesh (max. 2-3.5% Zr)
Flammable
Hafnium ≥99.6% (trace metals basis excluding Zr), powder -60+325 mesh (max. 2-3.5% Zr)
Catalog # AA10202-09
CAS Number:  
Flammable
Hafnium ≥99.6% (trace metals basis excluding Zr), powder -60+325 mesh (max. 2-3.5% Zr)
Catalog # AA10202-09
Supplier Number:  010202-09
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    10 g
  • Cat. No.:
    AA10202-09
  • Supplier no.:
    10202-09

Specifications

About this item

CAS Number:7440-58-6
 :
Formula:Hf
MDL Number:MFCD00011032
 :178.49
Merck Index:12,04618
Boiling Pt:5400 °C (1013 hPa)
Melting Pt:2224 °C
Density:13.31 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-166-4
UN:2545
ADR:4.2,II
 :GHS02